2-Chlor-4-nitrotoluol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23542.14
B23542.22
B23542.14EA
35.9
EUR
B23542.14
B23542.22
2-Chlor-4-nitrotoluol ≥98%
2-Chlor-4-nitrotoluol
Purity: 98%
CAS No.: 121-86-8
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Melting Point: 62-66°
Boiling Point: 260°
UN No.: 3457
MDL No.: MFCD00007210
Formel:
C₇H₆ClNO₂ Siedepunkt: 260 °C Schmelzpunkt: 62…66 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007210 CAS-Nummer: 121-86-8 EINECS: 204-501-7 UN-Nummer: 3457 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...