5-Nitro-o-toluylsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L11862.06
L11862.14
AB172032.0025
L11862.06EA
102
EUR
L11862.06
L11862.14
ABCRAB172032.0025
5-Nitro-o-toluylsäure ≥98%
5-Nitro-o-toluylsäure
Purity: 98+%
CAS No.: 1975-52-6
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 176-181°
Beilstein Reference: 2451300
Formel:
C₈H₇NO₄ Schmelzpunkt: 176…181 °C Dichte: 1,49 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007371 CAS-Nummer: 1975-52-6 EINECS: 217-829-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...