3-Brom-p-toluylsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A11496.06 A11496.14 A11496.22
A11496.06EA 42 EUR
A11496.06 A11496.14 A11496.22
3-Brom-p-toluylsäure ≥98%
3-Brom-p-toluylsäure

Purity: 98+% CAS No.: 7697-26-9 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Odour: Odourless Melting Point: 206-211° Beilstein Reference: 1936510 MDL No.: MFCD00002488

Formel: C₈H₇BrO₂
Schmelzpunkt: 206…211 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002488
CAS-Nummer: 7697-26-9
EINECS: 231-712-1
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR