1-Benzyloxy-4-iodbenzol ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-(Benzyloxy)-4-iodobenzene, 4-Benzyloxyiodobenzene

L16092.06 L16092.14
L16092.06EA 93.6 EUR
L16092.06 L16092.14
1-Benzyloxy-4-iodbenzol ≥98%
1-Benzyloxy-4-iodbenzol

Purity: 98+% CAS No.: 19578-68-8 Molecular Formula: C13H11IO Formula Weight: 310.14 Melting Point: 60-64° Boiling Point: 140°/1mm MDL No.: MFCD00079704

Formel: C₁₃H₁₁IO
Siedepunkt: 140 °C (1 mmHg)
Schmelzpunkt: 60…64 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00079704
CAS-Nummer: 19578-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR