6-Methoxy-2-pyridinboronsäurepinakolester ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Methoxy-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methoxypyridine-2-boronic acid pinacol ester
H27263.14
H27263.06
H27263.14EA
892
EUR
H27263.14
H27263.06
6-Methoxy-2-pyridinboronsäurepinakolester ≥97%
6-Methoxy-2-pyridinboronsäurepinakolester
Purity: 97%
CAS No.: 1034297-69-2
Molecular Formula: C12H18BNO3
Formula Weight: 235.09
Melting Point: 68-72°
MDL No.: MFCD06798266
Formel:
C₁₂H₁₈BNO₃ Schmelzpunkt: 68…72 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06798266 CAS-Nummer: 1034297-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...