1-Ethyl-3-methylimidazoliummethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethyl-3-methylimidazolium Methanesulphonate

29164-1KG-F 29164-100G-F
SIAL29164-1KG-FEA 941 EUR
SIAL29164-1KG-F SIAL29164-100G-F
1-Ethyl-3-methylimidazoliummethansulfonat, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium methanesulphonate
Formel: C₇H₁₄N₂O₃S
MDL Number: MFCD06798171
CAS-Nummer: 145022-45-3
UN-Nummer: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR