2-Fluorbenzothiazol

Lieferant: Thermo Scientific

AC33656.250 AC33656.1
MAYBAC33656.250EA 0 EUR
MAYBAC33656.250 MAYBAC33656.1
2-Fluorbenzothiazol
2-Fluorobenzothiazole
Formel: C₇H₄FNS
MDL Number: MFCD00047254
CAS-Nummer: 1123-98-4
EINECS: 214-384-4
UN-Nummer: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR