2-Chlor-6-fluorbenzothiazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chloro-6-fluorobenzo[d]thiazole
H32383.03
H32383.06
H32383.03EA
77.1
EUR
H32383.03
H32383.06
2-Chlor-6-fluorbenzothiazol ≥98%
2-Chloro-6-fluorobenzothiazole
Purity: 98%
CAS No.: 399-74-6
Molecular Formula: C7H3ClFNS
Formula Weight: 187.62
Melting Point: 89-93°
MDL No.: MFCD04448822
Formel:
C₇H₃ClFNS Schmelzpunkt: 89…93 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04448822 CAS-Nummer: 399-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...