2-(3-Chlor-4-fluorphenyl)indol

Lieferant: Thermo Scientific

SEW02771.1 SEW02771.10
MAYBSEW02771.1EA 0 EUR
MAYBSEW02771.1 MAYBSEW02771.10
2-(3-Chlor-4-fluorphenyl)indol
2-(3-Chloro-4-fluorophenyl)indole
Formel: C₁₄H₉ClFN
MDL Number: MFCD00068179
CAS-Nummer: 1868-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR