Ethyl-2-indolcarboxylat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Carbethoxyinole, Ethyl 1H-indole-2-carboxylate, Indole-2-Carboxylic Acid Ethyl Ester
M-Clarity™ quality level = MQ100
E32800-25G
E32800-5G
SIALE32800-25GEA
183
EUR
SIALE32800-25G
SIALE32800-5G
Ethyl-2-indolcarboxylat, Sigma-Aldrich®
Ethyl-2-indolcarboxylat
Formel:
C₁₁H₁₁NO₂ |
MDL Number:
MFCD00005609 CAS-Nummer: 3770-50-1 EINECS: 223-206-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...