5-Chloroxindol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-oxindole
A19210.06
A19210.14
A19210.06EA
183
EUR
A19210.06
A19210.14
5-Chloroxindol ≥98%
5-Chloroxindol
Purity: 98%
CAS No.: 17630-75-0
Molecular Formula: C8H6ClNO
Formula Weight: 167.60
Melting Point: 196-200°
MDL No.: MFCD00191927
Formel:
C₈H₆ClNO Molekulargewicht: 167.59 g/mol Schmelzpunkt: 196…200 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191927 CAS-Nummer: 17630-75-0 EINECS: 412-200-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...