1-Benzyl-4-piperidon 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone
106131000
106132500
ACRO106131000EA
90.9
EUR
ACRO106131000
ACRO106132500
1-Benzyl-4-piperidon 99%
1-Benzyl-4-piperidon
Formel:
C₁₂H₁₅NO Siedepunkt: 133…135 °C (7 mmHg) Dichte: 1.06 g/cm³ Flammpunkt: 71 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00006192 CAS-Nummer: 3612-20-2 EINECS: 222-782-4 |
Spezifikation Testergebnisse
Appearance | Clear colorless to straw yellow Viscous liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5390 to 1.5420 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥98.5 % |
Titration with HClO4 | 98 to 102 % |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤10 APHA |
Stabilizer | 50 to 100 ppm Amylene |
Residue after evaporation | ≤0.002 % |
Free halogens | Passes test |
Titrable acid | ≤0.0003 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...