1-Boc-4-piperidon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone
M-Clarity™ quality level = MQ100
461350-100G
461350-25G
461350-5G
SIAL461350-100GEA
471
EUR
SIAL461350-100G
SIAL461350-25G
SIAL461350-5G
1-Boc-4-piperidon, Sigma-Aldrich®
1-Boc-4-piperidon
Formel:
C₁₀H₁₇NO₃ |
MDL Number:
MFCD00151800 CAS-Nummer: 79099-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...