o-Dianisidin Dihydrochlorid ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3,3'-Dimethoxybenzidine dihydrochloride
A17175.03
A17175.06
A17175.03EA
35.8
EUR
A17175.03
A17175.06
o-Dianisidin Dihydrochlorid ≥99%
o-Dianisidin Dihydrochlorid
Purity: 99%
CAS No.: 20325-40-0
Molecular Formula: C14H16N2O2.2HCl
Formula Weight: 317.21
Melting Point: 268° dec.
Beilstein Reference: 3917996
Formel:
C₁₄H₁₈Cl₂N₂O₂ Schmelzpunkt: 268 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012488 CAS-Nummer: 20325-40-0 EINECS: 243-737-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...