o-Dianisidin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4,4'-Diamino-3,3'-dimethoxybiphenyl, 3,3'-Dimethoxybenzidine

A17150.06 A17150.14 A17150.22
A17150.06EA 36.7 EUR
A17150.06 A17150.14 A17150.22
o-Dianisidin ≥98%
o-Dianisidin

Purity: 98+% CAS No.: 119-90-4 Molecular Formula: C14H16N2O2 Formula Weight: 244.30 Flash Point: 206°(402°F) Melting Point: 134-138° Beilstein Reference: 1879884 Merck Reference: 14,2991 MDL No.: MFCD00008372

Formel: C₁₄H₁₆N₂O₂
Schmelzpunkt: 134…138 °C
Flammpunkt: 206 °C (402 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008372
CAS-Nummer: 119-90-4
EINECS: 204-355-4
Merck Index: 14,2991

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR