2-Chloraniliniumchlorid

Lieferant: Thermo Scientific

Synonyme: 2-Chloroaniline hydrochloride

SB01948.10 SB01948.50
MAYBSB01948.10EA 0 EUR
MAYBSB01948.10 MAYBSB01948.50
2-Chloraniliniumchlorid
2-Chloraniliniumchlorid
Formel: C₆H₇Cl₂N
MDL Number: MFCD00053282
CAS-Nummer: 137-04-2
EINECS: 205-274-7
UN-Nummer: 2018
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR