1-Ethyl-3-methylimidazoliumdiethylphosphat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-ethyl-3-methylimidazolium diethylphosphate

M-Clarity™ quality level = MQ100

671541-50G 671541-5G
SIAL671541-50GEA 292 EUR
SIAL671541-50G SIAL671541-5G
1-Ethyl-3-methylimidazoliumdiethylphosphat, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium diethylphosphate
Formel: C₁₀H₂₁N₂O₄P
MDL Number: MFCD09953486
CAS-Nummer: 848641-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR