1-Ethyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulphonyl)imide, [EMIIm], 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulphonyl)imide

711691-100G 711691-1KG
SIAL711691-100GEA 334 EUR
SIAL711691-100G SIAL711691-1KG
1-Ethyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumbis(trifluormethylsulfonyl)imid
Formel: C₈H₁₁F₆N₃O₄S₂
Molekulargewicht: 391.32 g/mol
MDL Number: MFCD03788927
CAS-Nummer: 174899-82-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR