3,5-Bis(trifluormethyl)thiophenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Bis(trifluoromethyl)benzenethiol

M-Clarity™ quality level = MQ200

536083-10ML 536083-1ML
SIAL536083-10MLEA 619 EUR
SIAL536083-10ML SIAL536083-1ML
3,5-Bis(trifluormethyl)thiophenol, Sigma-Aldrich®
3,5-Bis(trifluormethyl)thiophenol
Formel: C₈H₄F₆S
MDL Number: MFCD00042273
CAS-Nummer: 130783-02-7
UN-Nummer: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR