1-Acetyl-4-piperidon ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Acetyl-4-piperidone

A13512.06 A13512.14
A13512.06EA 49.7 EUR
A13512.06 A13512.14
1-Acetyl-4-piperidon ≥99%
1-Acetyl-4-piperidon

Purity: 99% CAS No.: 32161-06-1 Molecular Formula: C7H11NO2 Formula Weight: 141.17 Flash Point: 112°(234°F) Refractive Index: 1.5030 Boiling Point: 218° Beilstein Reference: 116031 MDL No.: MFCD00006190

Formel: C₇H₁₁NO₂
Siedepunkt: 218 °C
Dichte: 1.146
Flammpunkt: 112 °C (234 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006190
CAS-Nummer: 32161-06-1
EINECS: 250-937-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR