1-Methyl-3-octylimidazoliumchlorid, Sigma-Aldrich®
Lieferant: Merck
95803-50G
95803-5G
SIAL95803-50GEA
181
EUR
SIAL95803-50G
SIAL95803-5G
1-Methyl-3-octylimidazoliumchlorid, Sigma-Aldrich®
1-Methyl-3-octylimidazolium chloride
Formel:
C₁₂H₂₃ClN₂ |
MDL Number:
MFCD03095432 CAS-Nummer: 64697-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...