1-Methyl-3-octylimidazoliumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Methyl-3-n-octylimidazolium Tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, [OMIM] [BF4]
96324-50G
96324-5G
SIAL96324-50GEA
273
EUR
SIAL96324-50G
SIAL96324-5G
1-Methyl-3-octylimidazoliumtetrafluoroborat, Sigma-Aldrich®
1-Methyl-3-octylimidazolium tetrafluoroborate
Formel:
C₁₂H₂₃BF₄N₂ |
MDL Number:
MFCD03095434 CAS-Nummer: 244193-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...