Pyrogalloltrimethylether ≥98%
Lieferant: Thermo Fisher Scientific
A14572.14
A14572.22
A14572.36
A14572.14EA
31.9
EUR
A14572.14
A14572.22
A14572.36
Pyrogalloltrimethylether ≥98%
Pyrogalloltrimethylether
Purity: 98+%
CAS No.: 634-36-6
Molecular Formula: C9H12O3
Formula Weight: 168.19
Flash Point: 112°(233°F)
Melting Point: 43-45°
Boiling Point: 241°
Beilstein Reference: 1910422
MDL No.: MFCD00008358
Formel:
C₉H₁₂O₃ Siedepunkt: 241 °C Schmelzpunkt: 43…45 °C Dichte: 1,112 Flammpunkt: 112 °C (233 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008358 CAS-Nummer: 634-36-6 EINECS: 211-207-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...