(S)-(+)-2-Aminobuttersäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid
M-Clarity™ Qualitätsstufe = MQ400
A2536-1G
A2536-5G
SIALA2536-1GEA
154
EUR
SIALA2536-1G
SIALA2536-5G
(S)-(+)-2-Aminobuttersäure, Sigma-Aldrich®
(S)-(+)-2-Aminobuttersäure
Formel:
C₄H₉NO₂ Molekulargewicht: 103.12 g/mol |
MDL Number:
MFCD00064415 CAS-Nummer: 1492-24-6 EINECS: 216-083-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...