(R)-(-)-2-Aminobuttersäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-()-2-Aminobutyric acid, D-2-Aminobutyric acid, D(-)-2-Aminobutyric acid, H-D-Abu-OH

M-Clarity™ quality level = MQ100

116122-1G 116122-5G
SIAL116122-1GEA 27.2 EUR
SIAL116122-1G SIAL116122-5G
(R)-(-)-2-Aminobuttersäure, Sigma-Aldrich®
(R)-(-)-2-Aminobuttersäure
Formel: C₄H₉NO₂
MDL Number: MFCD00064414
CAS-Nummer: 2623-91-8
EINECS: 220-084-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR