(±)-2-Aminobuttersäure 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-DL-Abu-OH, DL-2-Aminobutyric acid, DL-α-Amino-N-Butyric Acid
L06035.14
L06035.22
L06035.14EA
47.5
EUR
L06035.14
L06035.22
(±)-2-Aminobuttersäure 99%
(±)-2-Aminobuttersäure
Purity: 99%
CAS No.: 2835-81-6
Molecular Formula: C4H9NO2
Formula Weight: 103.12
Melting Point: ca 291° dec.
Beilstein Reference: 635889
Merck Reference: 14,428
MDL No.: MFCD00008093
Formel:
C₄H₉NO₂ Schmelzpunkt: ∼ 291 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008093 CAS-Nummer: 2835-81-6 EINECS: 220-616-5 Merck Index: 14,428 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...