1-Methyl-4-piperidon ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N-Methyl-4-piperidone

A13352.0B A13352.18 A13352.22 A13352.30
A13352.0BEA 758 EUR
A13352.0B A13352.18 A13352.22 A13352.30
1-Methyl-4-piperidon ≥98%
1-Methyl-4-piperidon

Purity: 98% CAS No.: 1445-73-4 Molecular Formula: C6H11NO Formula Weight: 113.16 Storage Temperature: 2°C to 8°C Flash Point: 60°(140°F) Refractive Index: 1.4610 Odour: Amine-like pH: 12 Boiling Point: 55-58°/10mm Beilstein Reference: 106924 UN No.: 2733 MDL No.: MFCD00006191

Formel: C₆H₁₁NO
Siedepunkt: 55…58 °C (10 mmHg)
Dichte: 0.973
Flammpunkt: 60 °C (140 °F)
MDL Number: MFCD00006191
CAS-Nummer: 1445-73-4
EINECS: 215-895-5
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR