Ethyl-4-brombutyrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl 4-bromobutyrate
A13470.0B
A13470.18
A13470.30
A13470.0BEA
517
EUR
A13470.0B
A13470.18
A13470.30
Ethyl-4-brombutyrat ≥98%
Ethyl-4-brombutyrat
Purity: 98%
CAS No.: 2969-81-5
Molecular Formula: Br(CH2)3CO2CH2CH3
Formula Weight: 195.06
Flash Point: 90°(194°F)
Physical Form: Liquid
Density: 1.363
Refractive Index: 1.4560
Appearance: Colourless to pale yellow
Odour: Odourless
Boiling Point: 80-82°/10mm
Beilstein Reference: 1749700
MDL No.: MFCD00000259
Formel:
C₆H₁₁BrO₂ Siedepunkt: 80…82 °C (10 mmHg) Dichte: 1.363 Flammpunkt: 90 °C (194 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00000259 CAS-Nummer: 2969-81-5 EINECS: 221-005-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...