Magnesiumsilikat 60-120 mesh für die Chromatographie
Lieferant: Thermo Fisher Scientific
Synonyme: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
B22490.0B B22490.30
B22490.0BEA 301 EUR
B22490.0B B22490.30
Magnesiumsilikat 60-120 mesh für die Chromatographie
Magnesiumsilikat
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.
Notes: Incompatible with strong acids and hydrogen fluoride.
Formel: MgO₃Si
Molekulargewicht: 100.38 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00078249
CAS-Nummer: 1343-88-0
EINECS: 215-681-1