Guajacol zur Synthese, Sigma-Aldrich®
Lieferant: Merck
8.18339.0005
8.18339.0250
8.18339.1000
8.18339.0005EA
28
EUR
8.18339.0005
8.18339.0250
8.18339.1000
Guajacol zur Synthese, Sigma-Aldrich®
Guajacol
Spezifikation Testergebnisse
Solubility | 10 g/l (30 °C) |
Melting Point | 28 - 32 °C |
Molar Mass | 124.14 g/mol |
Boiling Point | 205 °C |
Vapor Pressure | 0.1 hPa (25 °C) |
Flash Point | 82 °C |
Density | 1.13 g/cm3 (21 °C) |
pH | 5.4 (10 g/l, H2O, 20 °C) |
Ignition Point | 750 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...