Guajacol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A16319.0B A16319.0I A16319.30
A16319.0BEA 115 EUR
A16319.0B A16319.0I A16319.30
Guajacol ≥98%
Guajacol

Purity: 98+% CAS No.: 90-05-1 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 82°(179°F) Physical Form: Solid Density: 1.129 Refractive Index: 1.5430 Sensitivity: Air and Light Sensitive Appearance: White to pale brown Odour: Characteristic pH: 5.4 Melting Point: 28-31° Boiling Point: 204-206° Beilstein Reference: 508112 Merck Reference: 14,4553 MDL No.: MFCD00002185

Formel: C₇H₈O₂
Siedepunkt: 204…206 °C
Schmelzpunkt: 28…31 °C
Dichte: 1,129
Flammpunkt: 82 °C (179 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002185
CAS-Nummer: 90-05-1
EINECS: 201-964-7
UN-Nummer: 2810
ADR: 6.1,III
Merck Index: 14,4553

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR