4-Iodanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B21936.18 B21936.30
B21936.18EA 58.7 EUR
B21936.18 B21936.30
4-Iodanilin ≥98%
4-Iodanilin

Purity: 98% CAS No.: 540-37-4 Molecular Formula: C6H6IN Formula Weight: 219.03 Melting Point: 61-64° Beilstein Reference: 774101 Merck Reference: 14,5027 MDL No.: MFCD00007848

Formel: C₆H₆IN
Siedepunkt: 262 °C (1013 hPa)
Schmelzpunkt: 61…64 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007848
CAS-Nummer: 540-37-4
EINECS: 208-743-4
Merck Index: 14,5027

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR