1,2-Difluor-4-iodbenzol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A11339.06 A11339.14
A11339.06EA 37.5 EUR
A11339.06 A11339.14
1,2-Difluor-4-iodbenzol ≥99%

Purity: 99% CAS No.: 64248-58-4 Molecular Formula: C6H3F2I Formula Weight: 239.99 Flash Point: 72°(161°F) Refractive Index: 1.5590 Boiling Point: 177-178° Beilstein Reference: 2081099 MDL No.: MFCD00044630

Formel: C₆H₃F₂I
Siedepunkt: 177…178 °C
Dichte: 1,990
Flammpunkt: 72 °C (161 °F)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00044630
CAS-Nummer: 64248-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR