3,5-Dichlorthiophenol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,5-Dichlorobenzene-1-thiol, 3,5-Dichlorobenzenethiol, 1,3-Dichloro-5-mercaptobenzene

L10697.03 L10697.06
L10697.03EA 50 EUR
L10697.03 L10697.06
3,5-Dichlorthiophenol ≥97%
3,5-Dichlorthiophenol

Purity: 97% CAS No.: 17231-94-6 Molecular Formula: C6H4Cl2S Formula Weight: 179.07 Odour: Unpleasant Melting Point: 60-63° Boiling Point: 270-272° Beilstein Reference: 2041184 MDL No.: MFCD00041423

Formel: C₆H₄Cl₂S
Siedepunkt: 270…272 °C
Schmelzpunkt: 60…63 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00041423
CAS-Nummer: 17231-94-6
UN-Nummer: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR