Precision balances, Excellence XA Series
Lieferant: Mettler - Toledo
11126810 11126815 11126825 11126820 11126830 11126835
611-2474EA 0 EUR
611-2474 611-2475 611-2477 611-2476 611-2478 611-2479
Precision balances, Excellence XA Series
Waagen
The most straighforward series in the Excellence range, the XA precision balances are simple to use and provide fast, reliable, accurate weighing using MonoBloc HighSpeed technology. Draught shields are standard on the small square platform size (see models denoted * in the table). The monochrome touch screen provides fast and easy access to built-in applications.
  • Durable, robust design with stainless steel pan
  • FACT fully automatic temperature controlled internal adjustment
  • Chemically resistant housing and overload protection for everyday use in difficult weighing environments
  • Intuitive interface with data input via alphanumeric touch pad
  • Easy to clean, with removable terminal
*Includes draught shield.
Bestellinformation: For details of the full range of accessories for Excellence balances please contact the Avantor Technical Office on 01342 337447.