3-Chloranisol ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
1-Chloro-3-methoxybenzene
A17327.18
A17327.30
A17327.18EA
63.9
EUR
A17327.18
A17327.30
3-Chloranisol ≥98%
3-Chloranisol
Purity: 98+%
CAS No.: 2845-89-8
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Flash Point: 73°(163°F)
Refractive Index: 1.5360
Boiling Point: 192-194°
Beilstein Reference: 2041497
MDL No.: MFCD00000591
Formel:
C₇H₇ClO Siedepunkt: 192…194 °C Dichte: 1.168 Flammpunkt: 73 °C (163 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00000591 CAS-Nummer: 2845-89-8 EINECS: 220-642-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...