2,5-Dichloranisol ≥98%
Lieferant: Thermo Fisher Scientific
L13497.06
L13497.14
L13497.06EA
43.2
EUR
L13497.06
L13497.14
2,5-Dichloranisol ≥98%
2,5-Dichloranisol
Purity: 98%
CAS No.: 1984-58-3
Molecular Formula: C7H6Cl2O
Formula Weight: 177.03
Refractive Index: 1.5630
Boiling Point: 128-132°/3.5mm
MDL No.: MFCD00061123
Formel:
C₇H₆Cl₂O Siedepunkt: 128…132 °C (3,5 mmHg) Schmelzpunkt: 24 °C Dichte: 1,330 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00061123 CAS-Nummer: 1984-58-3 EINECS: 217-852-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...