2-Chlor-4-nitrophenol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B21561.09 B21561.18 B21561.30
B21561.09EA 37.3 EUR
B21561.09 B21561.18 B21561.30
2-Chlor-4-nitrophenol ≥97%
2-Chlor-4-nitrophenol

Purity: 97% CAS No.: 619-08-9 Molecular Formula: C6H4ClNO3 Formula Weight: 173.56 Odour: Characteristic Melting Point: 108-112° Beilstein Reference: 2046372 MDL No.: MFCD00043910

Formel: C₆H₄ClNO₃
Schmelzpunkt: 108…112 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00043910
CAS-Nummer: 619-08-9
EINECS: 210-578-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR