Multispectral source, BioLite™
Lieferant: UVP ULTRA VIOLET PRODUCTS
98-0072-08 98-0072-11
UVPA98-0072-08EA 7470 EUR
UVPA98-0072-08 UVPA98-0072-11
Multispectral source, BioLite™
Beleuchtung Spektrallampen
The BioLite MultiSpectral Source uses a 150 W quartz halogen or 150 W xenon light source with fibre optics to tightly control the output spectrum.
  • Xenon light source is strongly recommended for customers with NIR requirement
A six position dial allows selection of the light intensity output for different applications.
* No excitation filters are included with the BioLite unit. Please ask for details of the available choice.