Precision balances, NewClassic, MS series
Lieferant: Mettler - Toledo
11124516 11124489 11124522 11124531 11124540 11124564 11124611 11124629 11124635 11124662 11124492 11124498 11124561 11124504 11124528 11124537 MS6001S
611-2486EA 0 EUR
611-2486 611-2482 611-2487 611-2489 611-2491 611-2493 611-2494 611-2495 611-2496 611-2497 611-2483 611-2484 611-2492 611-2485 611-2488 611-2490 METBMS6001S
Precision balances, NewClassic, MS series
Waagen
A range of classic design, precision balances that incorporates new technology to deliver rugged, reliable, effective performance. Applications include formulation, statistics, checkweighing, piece counting, dynamic weighing and totalling.
  • MonoBloc weighing technology for fast, repeatable results
  • High grade chemically-resistant, die-cast aluminium housing
  • USB connectivity to PC for easy transfer of results
  • Diagnostics check helps audit the balance adjustment history
  • FACT internal adjustment except models denoted with suffix E which require manual calibration
  • One-touch function via user-defined Smart Keys
Three figure balances with draught shield
QuickLock - for rapid and comfortable dismantling of the draught shield without tools.
Glass panels are dishwasher-safe.
Bestellinformation: OIML certified versions for all models in tables below, together with a full range of accessories are also available - please see the Avantor website or contact your local Avantor sales office.