2-Nitrobenzoesäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

A10393.0E A10393.22 A10393.36
A10393.0EEA 332 EUR
A10393.0E A10393.22 A10393.36
2-Nitrobenzoesäure 95%
2-Nitrobenzoesäure

Purity: 95% CAS No.: 552-16-9 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 146-149° Beilstein Reference: 1910632 Merck Reference: 14,6589 MDL No.: MFCD00007137

Formel: C₇H₅NO₄
Molekulargewicht: 167.12 g/mol
Schmelzpunkt: 146…149 °C
Dichte: 1.58
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007137
CAS-Nummer: 552-16-9
EINECS: 209-004-9
Merck Index: 12,06686

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR