5-Nitrosalicylaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Hydroxy-5-nitrobenzaldehyde

A10264.06 A10264.14 A10264.22
A10264.06EA 57.2 EUR
A10264.06 A10264.14 A10264.22
5-Nitrosalicylaldehyd ≥98%
5-Nitrosalicylaldehyd

Purity: 98% CAS No.: 97-51-8 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 123-127° Beilstein Reference: 512565

Formel: C₇H₅NO₄
Schmelzpunkt: 123…127 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007337
CAS-Nummer: 97-51-8
EINECS: 202-587-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR