4-Brom-2-fluorphenol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Bromo-3-fluoro-4-hydroxybenzene
A10804.14
A10804.06
A10804.22
A10804.14EA
76.2
EUR
A10804.14
A10804.06
A10804.22
4-Brom-2-fluorphenol ≥97%
4-Brom-2-fluorphenol
Purity: 97+%
CAS No.: 2105-94-4
Molecular Formula: C6H4BrFO
Formula Weight: 191.00
Flash Point: 98°(208°F)
Refractive Index: 1.5660
Boiling Point: 82-83°/10mm
Beilstein Reference: 1861281
MDL No.: MFCD00011722
Formel:
C₆H₄BrFO Siedepunkt: 82…83 °C (10 mmHg) Dichte: 1.744 Flammpunkt: 98 °C (208 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011722 CAS-Nummer: 2105-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...