L-Cycloserin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

C1159-25MG C1159-100MG C1159-10MG C1159-250MG
SIALC1159-25MGEA 68.3 EUR
SIALC1159-25MG SIALC1159-100MG SIALC1159-10MG SIALC1159-250MG
L-Cycloserin, Sigma-Aldrich®
L-Cycloserin
Formel: C₃H₆N₂O₂
MDL Number: MFCD00064324
CAS-Nummer: 339-72-0
EINECS: 206-427-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR