4-Iodphenylhydrazin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
638463-1G
638463-5G
SIAL638463-1GEA
88.6
EUR
SIAL638463-1G
SIAL638463-5G
4-Iodphenylhydrazin, Sigma-Aldrich®
4-Iodphenylhydrazin
Formel:
C₆H₇IN₂ |
MDL Number:
MFCD00052688 CAS-Nummer: 13116-27-3 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...