2-Ethylhexylmethacrylat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methacrylic acid 2-ethylhexyl ester
M-Clarity™ Qualitätsstufe = MQ200
290807-1L
290807-25ML
SIAL290807-1LEA
229
EUR
SIAL290807-1L
SIAL290807-25ML
2-Ethylhexylmethacrylat, Sigma-Aldrich®
2-Ethylhexylmethacrylat
Formel:
C₁₂H₂₂O₂ Molekulargewicht: 198.31 g/mol |
MDL Number:
MFCD00009494 CAS-Nummer: 688-84-6 EINECS: 211-708-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...