Bis(4-chlorphenyl)sulfon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4,4'-Dichloro diphenyl sulphone, 4-Chlorophenyl sulphone, 4,4'-sulphonylbis(chlorobenzene)
M-Clarity™ quality level = MQ100
151378-100G
151378-500G
SIAL151378-100GEA
66.8
EUR
SIAL151378-100G
SIAL151378-500G
Bis(4-chlorphenyl)sulfon, Sigma-Aldrich®
Bis(4-chlorphenyl)sulfon
Formel:
C₁₂H₈Cl₂O₂S |
MDL Number:
MFCD00000619 CAS-Nummer: 80-07-9 EINECS: 201-247-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...