Dapson, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
M-Clarity™ quality level = MQ200
8.21073.0005
8.21073.0250
8.21073.0005EA
25.9
EUR
8.21073.0005
8.21073.0250
Dapson, Sigma-Aldrich®
Dapson
Formel:
C₁₂H₁₂N₂O₂S |
MDL Number:
MFCD00007887 CAS-Nummer: 80-08-0 EINECS: 201-248-4 |
Spezifikation Testergebnisse
Solubility | 0.38 g/l (37 °C) |
Melting Point | 172 - 175 °C |
Molar Mass | 248.30 g/mol |
Bulk Density | 250 - 350 kg/m3 |
pH | 5.5 - 7.5 (H2O, 20 °C) (saturated aqueous solution) |
Synonyms | DDS, 4,4'-Sulfonyldianiline, Bis(p-aminophenyl) sulfone |
Hill Formula | C12H12N2O2S |
Chemical Formula | [4-(NH2)C6H4]2SO2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...