3-Fluorphenylhydraziniumchlorid

Lieferant: Thermo Scientific

Synonyme: (3-Fluorphenyl)hydrazin HCl, (3-Fluorophenyl)hydrazine hydrochloride, 3-Fluorophenylhydrazine hydrochloride, 1-(3-Fluorophenyl)hydrazine hydrochloride

SB01661.1 SB01661.10 SB01661.50
MAYBSB01661.1EA 0 EUR
MAYBSB01661.1 MAYBSB01661.10 MAYBSB01661.50
3-Fluorphenylhydraziniumchlorid
3-Fluorphenylhydraziniumchlorid
Formel: C₆H₈ClFN₂
MDL Number: MFCD00012934
CAS-Nummer: 2924-16-5
EINECS: 220-886-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR