2,4-Difluorphenylhydrazin

Lieferant: Thermo Scientific

RF01198.1 RF01198.10
MAYBRF01198.1EA 0 EUR
MAYBRF01198.1 MAYBRF01198.10
2,4-Difluorphenylhydrazin
2,4-Difluorphenylhydrazin
Formel: C₆H₆F₂N₂
MDL Number: MFCD00042256
CAS-Nummer: 40594-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR